Assay Details
Gene Symbol: Erbb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017218.3 | NP_058914.2 | 22 - 23 | 2829 | 70 | |
GenBank mRNA | U29339.2 | - | 22 - 23 | 2829 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
