Assay Details
Gene Symbol: Nabp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244819.1 | NP_001231748.1 | 1 - 2 | 95 | 128 | |
GenBank mRNA | DV214819.1 | - | 1 - 2 | 95 | 128 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001244819.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
