Assay Details
Gene Symbol: Ptges3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130989.1 | NP_001124461.1 | 2 - 3 | 410 | 187 | |
XM_008765026.1 | XP_008763248.1 | 2 - 3 | 447 | 187 | ||
GenBank mRNA | BC166579.1 | - | 2 - 3 | 410 | 187 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
