Assay Details
Gene Symbol: G6pd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017006.2 | NP_058702.1 | 8 - 9 | 974 | 103 | |
GenBank mRNA | BC081820.1 | - | 8 - 9 | 887 | 103 | |
X07467.1 | - | 8 - 9 | 900 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
