Assay Details
Gene Symbol: Ikbkg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199103.1 | NP_954534.1 | 1 - 2 | 184 | 72 | |
XM_006229580.2 | XP_006229642.1 | 3 - 4 | 1432 | 72 | ||
XM_006229584.2 | XP_006229646.1 | 3 - 4 | 418 | 72 | ||
XM_006229585.2 | XP_006229647.1 | 2 - 3 | 378 | 72 | ||
XM_008773600.1 | XP_008771822.1 | 2 - 3 | 4489 | 72 | ||
XM_008773601.1 | XP_008771823.1 | 3 - 4 | 1432 | 72 | ||
XM_008773602.1 | XP_008771824.1 | 3 - 4 | 1446 | 72 | ||
XM_008773603.1 | XP_008771825.1 | 3 - 4 | 1437 | 72 | ||
XM_008773604.1 | XP_008771826.1 | 2 - 3 | 4489 | 72 | ||
XM_008773605.1 | XP_008771827.1 | 2 - 3 | 4489 | 72 | ||
XM_008773606.1 | XP_008771828.1 | 2 - 3 | 4489 | 72 | ||
XM_008773607.1 | XP_008771829.1 | 3 - 4 | 4529 | 72 | ||
XM_008773608.1 | XP_008771830.1 | 3 - 4 | 419 | 72 | ||
XM_008773609.1 | XP_008771831.1 | 3 - 4 | 627 | 72 | ||
XM_008773610.1 | XP_008771832.1 | 2 - 3 | 379 | 72 | ||
GenBank mRNA | AY392762.1 | - | 1 - 2 | 184 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
