Assay Details
Gene Symbol: Idh3g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031551.1 | NP_113739.1 | 4 - 5 | 295 | 79 | |
XM_006229551.1 | XP_006229613.1 | 2 - 3 | 717 | 79 | ||
XM_006229552.2 | XP_006229614.1 | 3 - 4 | 300 | 79 | ||
XM_006229553.2 | XP_006229615.1 | 4 - 5 | 320 | 79 | ||
XM_006229554.1 | XP_006229616.1 | 3 - 4 | 247 | 79 | ||
GenBank mRNA | BC083688.1 | - | 4 - 5 | 295 | 79 | |
FQ213717.1 | - | 4 - 5 | 174 | 79 | ||
FQ214482.1 | - | 4 - 5 | 261 | 79 | ||
FQ214526.1 | - | 4 - 5 | 261 | 79 | ||
FQ214664.1 | - | 4 - 5 | 236 | 79 | ||
FQ214718.1 | - | 4 - 5 | 311 | 79 | ||
FQ214896.1 | - | 4 - 5 | 246 | 79 | ||
FQ216691.1 | - | 4 - 5 | 251 | 79 | ||
FQ226989.1 | - | 3 - 4 | 173 | 79 | ||
X74125.1 | - | 4 - 5 | 221 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
