Assay Details
Gene Symbol: Idh3g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031551.1 | NP_113739.1 | 6 - 7 | 463 | 65 | |
XM_006229551.1 | XP_006229613.1 | 4 - 5 | 885 | 65 | ||
XM_006229552.2 | XP_006229614.1 | 5 - 6 | 468 | 65 | ||
XM_006229553.2 | XP_006229615.1 | 6 - 7 | 488 | 65 | ||
XM_006229554.1 | XP_006229616.1 | 5 - 6 | 415 | 65 | ||
GenBank mRNA | BC083688.1 | - | 6 - 7 | 463 | 65 | |
FQ213717.1 | - | 6 - 7 | 342 | 65 | ||
FQ214482.1 | - | 6 - 7 | 429 | 65 | ||
FQ214526.1 | - | 6 - 7 | 429 | 65 | ||
FQ214664.1 | - | 6 - 7 | 404 | 65 | ||
FQ214718.1 | - | 6 - 7 | 479 | 65 | ||
FQ214896.1 | - | 6 - 7 | 414 | 65 | ||
FQ226989.1 | - | 5 - 6 | 341 | 65 | ||
X74125.1 | - | 6 - 7 | 389 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
