Assay Details
Gene Symbol: Idh3g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031551.1 | NP_113739.1 | 7 - 8 | 598 | 103 | |
XM_006229551.1 | XP_006229613.1 | 5 - 6 | 1020 | 103 | ||
XM_006229552.2 | XP_006229614.1 | 6 - 7 | 603 | 103 | ||
XM_006229553.2 | XP_006229615.1 | 7 - 8 | 623 | 103 | ||
XM_006229554.1 | XP_006229616.1 | 6 - 7 | 550 | 103 | ||
GenBank mRNA | BC083688.1 | - | 7 - 8 | 598 | 103 | |
FQ213717.1 | - | 7 - 8 | 477 | 103 | ||
FQ214482.1 | - | 7 - 8 | 564 | 103 | ||
FQ214526.1 | - | 7 - 8 | 564 | 103 | ||
FQ214664.1 | - | 7 - 8 | 539 | 103 | ||
FQ214718.1 | - | 7 - 8 | 615 | 103 | ||
FQ214896.1 | - | 7 - 8 | 549 | 103 | ||
FQ216691.1 | - | 7 - 8 | 554 | 103 | ||
FQ226989.1 | - | 6 - 7 | 476 | 103 | ||
X74125.1 | - | 7 - 8 | 524 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
