Assay Details
Gene Symbol: Idh3g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006229551.1 | XP_006229613.1 | 1 - 2 | 81 | 126 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006229551.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
