Assay Details
Gene Symbol: Fam3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109324.1 | NP_001102794.1 | 1 - 2 | 337 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001109324.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
