Assay Details
Gene Symbol: Fam3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109324.1 | NP_001102794.1 | 2 - 3 | 410 | 74 | |
XM_006229603.2 | XP_006229665.1 | 1 - 2 | 567 | 74 | ||
XM_006229604.2 | XP_006229666.1 | 1 - 2 | 569 | 74 | ||
XM_006229605.2 | XP_006229667.1 | 1 - 2 | 568 | 74 | ||
GenBank mRNA | BC169081.1 | - | 1 - 2 | 474 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC169081.1, NM_001109324.1, XM_006229603.2, XM_006229604.2, XM_006229605.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
