Assay Details
Gene Symbol: Scgn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008771653.1 | XP_008769875.1 | 8 - 9 | 931 | 130 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
