Assay Details
Gene Symbol: Nde1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053347.1 | NP_445799.1 | 1 - 2 | 79 | 113 | |
XM_006245849.2 | XP_006245911.1 | 2 - 3 | 196 | 113 | ||
XM_006245850.2 | XP_006245912.1 | 3 - 4 | 228 | 113 | ||
XM_006245851.1 | XP_006245913.1 | 2 - 3 | 196 | 113 | ||
XM_008767443.1 | XP_008765665.1 | 2 - 3 | 180 | 113 | ||
XM_008767444.1 | XP_008765666.1 | 2 - 3 | 180 | 113 | ||
XM_008767445.1 | XP_008765667.1 | 2 - 3 | 180 | 113 | ||
GenBank mRNA | AF240463.1 | - | 1 - 2 | 79 | 113 | |
BC081759.1 | - | 1 - 2 | 91 | 113 |
Gene Symbol: LOC103690118 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006245690.2 | XP_006245752.1 | 2 - 3 | 225 | 113 | |
XM_008767446.1 | XP_008765668.1 | 3 - 4 | 266 | 113 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
