Assay Details
Gene Symbol: Nde1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053347.1 | NP_445799.1 | 5 - 6 | 705 | 105 | |
XM_006245849.2 | XP_006245911.1 | 6 - 7 | 822 | 105 | ||
XM_006245850.2 | XP_006245912.1 | 7 - 8 | 854 | 105 | ||
XM_006245851.1 | XP_006245913.1 | 6 - 7 | 822 | 105 | ||
XM_008767443.1 | XP_008765665.1 | 7 - 8 | 875 | 105 | ||
XM_008767444.1 | XP_008765666.1 | 7 - 8 | 875 | 105 | ||
XM_008767445.1 | XP_008765667.1 | 7 - 8 | 875 | 105 | ||
GenBank mRNA | AF240463.1 | - | 5 - 6 | 705 | 105 | |
BC081759.1 | - | 5 - 6 | 717 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
