Assay Details
Gene Symbol: Anxa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019905.1 | NP_063970.1 | 10 - 11 | 823 | 68 | |
GenBank mRNA | BC059136.1 | - | 10 - 11 | 842 | 68 | |
FQ225322.1 | - | 10 - 11 | 841 | 68 | ||
FQ225353.1 | - | 7 - 8 | 611 | 68 | ||
L13039.1 | - | 9 - 10 | 794 | 68 | ||
S73559.1 | - | 10 - 11 | 825 | 68 | ||
X66871.1 | - | 10 - 11 | 823 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
