Assay Details
Gene Symbol: Anxa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019905.1 | NP_063970.1 | 5 - 6 | 401 | 81 | |
GenBank mRNA | BC059136.1 | - | 5 - 6 | 420 | 81 | |
FQ225322.1 | - | 5 - 6 | 417 | 81 | ||
L13039.1 | - | 4 - 5 | 372 | 81 | ||
S73559.1 | - | 5 - 6 | 403 | 81 | ||
X66871.1 | - | 5 - 6 | 401 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
