Assay Details
Gene Symbol: Anxa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019905.1 | NP_063970.1 | 8 - 9 | 633 | 69 | |
GenBank mRNA | BC059136.1 | - | 8 - 9 | 652 | 69 | |
FQ219918.1 | - | 3 - 4 | 199 | 69 | ||
FQ225353.1 | - | 5 - 6 | 421 | 69 | ||
L13039.1 | - | 7 - 8 | 604 | 69 | ||
S73559.1 | - | 8 - 9 | 635 | 69 | ||
X66871.1 | - | 8 - 9 | 633 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
