Assay Details
Gene Symbol: Bnip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106835.1 | NP_001100305.1 | 6 - 7 | 827 | 105 | |
XM_006243370.2 | XP_006243432.1 | 6 - 7 | 871 | 105 | ||
XM_006243371.2 | XP_006243433.1 | 6 - 7 | 869 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
