Assay Details
Gene Symbol: Gtf2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006243380.2 | XP_006243442.1 | 1 - 2 | 106 | 108 | |
GenBank mRNA | BC168727.1 | - | 1 - 2 | 92 | 108 | |
FQ209546.1 | - | 111 | 108 | |||
FQ212213.1 | - | 1 - 2 | 91 | 108 | ||
FQ221947.1 | - | 1 - 2 | 97 | 108 | ||
FQ228685.1 | - | 1 - 2 | 111 | 108 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC168727.1, XM_006243380.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
