Assay Details
Gene Symbol: Gtf2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053345.1 | NP_445797.1 | 1 - 2 | 79 | 121 | |
XM_006243380.2 | XP_006243442.1 | 2 - 3 | 229 | 121 | ||
GenBank mRNA | AF000944.1 | - | 1 - 2 | 79 | 121 | |
BC168727.1 | - | 2 - 3 | 215 | 121 | ||
FQ209546.1 | - | 234 | 121 | |||
FQ212213.1 | - | 2 - 3 | 214 | 121 | ||
FQ221947.1 | - | 2 - 3 | 220 | 121 | ||
FQ228685.1 | - | 2 - 3 | 234 | 121 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
