Assay Details
Gene Symbol: Cdc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108352.1 | NP_001101822.1 | 5 - 6 | 598 | 100 | |
XM_006250570.2 | XP_006250632.1 | 4 - 5 | 430 | 100 | ||
XM_008769940.1 | XP_008768162.1 | 3 - 4 | 311 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
