Assay Details
Gene Symbol: Zfp644 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001139484.1 | NP_001132956.1 | 6 - 7 | 2918 | 126 | |
XM_008769932.1 | XP_008768154.1 | 2 - 3 | 2509 | 126 | ||
XM_008769934.1 | XP_008768156.1 | 2 - 3 | 2583 | 126 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
