Assay Details
Gene Symbol: Zfp644 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001139484.1 | NP_001132956.1 | 8 - 9 | 4237 | 106 | |
XM_006250550.1 | XP_006250612.1 | 3 - 4 | 3924 | 106 | ||
XM_006250554.2 | XP_006250616.1 | 3 - 4 | 3934 | 106 | ||
XM_008769931.1 | XP_008768153.1 | 3 - 4 | 3998 | 106 | ||
XM_008769932.1 | XP_008768154.1 | 4 - 5 | 3828 | 106 | ||
XM_008769934.1 | XP_008768156.1 | 4 - 5 | 3902 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
