Assay Details
Gene Symbol: Pknox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271278.1 | NP_001258207.1 | 7 - 8 | 920 | 89 | |
NM_001271279.1 | NP_001258208.1 | 8 - 9 | 1052 | 89 | ||
GenBank mRNA | FQ221048.1 | - | 8 - 9 | 1052 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
