Assay Details
Gene Symbol: Ahcy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017201.1 | NP_058897.1 | 4 - 5 | 468 | 119 | |
GenBank mRNA | M15185.1 | - | 4 - 5 | 468 | 119 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
