Assay Details
Gene Symbol: Nlrc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008772331.1 | XP_008770553.1 | 9 - 10 | 3847 | 64 |
Gene Symbol: LOC102553150 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006255134.2 | XP_006255196.1 | 2 - 3 | 772 | 64 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
