Assay Details
Gene Symbol: Got2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013177.2 | NP_037309.1 | 1 - 2 | 204 | 110 | |
GenBank mRNA | BC061792.1 | - | 1 - 2 | 152 | 110 | |
FM056810.1 | - | 1 - 2 | 204 | 110 | ||
M18467.1 | - | 1 - 2 | 175 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
