Assay Details
Gene Symbol: Riok3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108423.1 | NP_001101893.1 | 6 - 7 | 1599 | 128 | |
GenBank mRNA | FQ230531.1 | - | 3 - 4 | 375 | 128 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
