Assay Details
Gene Symbol: Gtf2h4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212501.2 | NP_997666.1 | 1 - 2 | 167 | 103 | |
GenBank mRNA | BC083703.1 | - | 1 - 2 | 125 | 103 | |
BC127469.1 | - | 1 - 2 | 139 | 103 | ||
CK472761.1 | - | 1 - 2 | 167 | 103 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC127469.1, NM_212501.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
