Assay Details
Gene Symbol: Abcf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109883.2 | NP_001103353.1 | 15 - 16 | 1495 | 67 | |
GenBank mRNA | AF293383.1 | - | 14 - 15 | 1391 | 67 | |
CB801303.1 | - | 3 - 4 | 308 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
