Assay Details
Gene Symbol: Abcf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109883.2 | NP_001103353.1 | 16 - 17 | 1617 | 71 | |
GenBank mRNA | AF293383.1 | - | 15 - 16 | 1513 | 71 | |
CV114142.1 | - | 1 - 2 | 47 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
