Assay Details
Gene Symbol: Gnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212500.3 | NP_997665.1 | 3 - 4 | 508 | 112 | |
GenBank mRNA | BC129081.1 | - | 3 - 4 | 413 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
