Assay Details
Gene Symbol: Gnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212500.3 | NP_997665.1 | 4 - 5 | 660 | 95 | |
GenBank mRNA | BC083888.1 | - | 1 - 2 | 153 | 95 | |
BC129081.1 | - | 4 - 5 | 565 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
