Assay Details
Gene Symbol: Cacnb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012828.2 | NP_036960.1 | 11 - 12 | 1080 | 116 | |
XM_006257329.2 | XP_006257391.1 | 11 - 12 | 1152 | 116 | ||
XM_006257330.2 | XP_006257392.1 | 11 - 12 | 1561 | 116 | ||
GenBank mRNA | M88751.1 | - | 11 - 12 | 1084 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
