Assay Details
Gene Symbol: Cacnb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012828.2 | NP_036960.1 | 6 - 7 | 583 | 106 | |
XM_006257329.2 | XP_006257391.1 | 6 - 7 | 655 | 106 | ||
XM_006257330.2 | XP_006257392.1 | 6 - 7 | 1064 | 106 | ||
GenBank mRNA | M88751.1 | - | 6 - 7 | 587 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
