Assay Details
Gene Symbol: Kansl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008765691.1 | XP_008763913.1 | 1 - 2 | 325 | 69 | |
GenBank mRNA | BC169120.1 | - | 1 - 2 | 355 | 69 |
Gene Symbol: LOC103690317 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008765853.1 | XP_008764075.1 | 325 | 69 | ||
XM_008765856.1 | XP_008764078.1 | 400 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008765691.1, XM_008765853.1, XM_008765856.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
