Assay Details
Gene Symbol: Ppp1r1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022676.3 | NP_073167.1 | 4 - 5 | 381 | 70 | |
XM_006242437.2 | XP_006242499.1 | 4 - 5 | 393 | 70 | ||
GenBank mRNA | AJ276593.1 | - | 4 - 5 | 254 | 70 | |
AY648296.1 | - | 4 - 5 | 381 | 70 | ||
BC078820.1 | - | 4 - 5 | 382 | 70 | ||
FQ217772.1 | - | 4 - 5 | 350 | 70 | ||
J05592.1 | - | 4 - 5 | 253 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
