Assay Details
Gene Symbol: Acvrl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022441.2 | NP_071886.1 | 3 - 4 | 462 | 113 | |
XM_006242303.2 | XP_006242365.1 | 4 - 5 | 863 | 113 | ||
XM_006242304.2 | XP_006242366.1 | 4 - 5 | 1467 | 113 | ||
XM_006242305.2 | XP_006242367.1 | 3 - 4 | 1482 | 113 | ||
XM_006242307.2 | XP_006242369.1 | 3 - 4 | 1352 | 113 | ||
XM_008765673.1 | XP_008763895.1 | 4 - 5 | 1526 | 113 | ||
GenBank mRNA | BC083173.1 | - | 3 - 4 | 462 | 113 | |
L36088.1 | - | 4 - 5 | 877 | 113 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
