Assay Details
Gene Symbol: Krt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008806.1 | NP_001008806.1 | 5 - 6 | 1029 | 104 | |
XM_006242395.2 | XP_006242457.1 | 5 - 6 | 1096 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
