Assay Details
Gene Symbol: Asic1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024154.2 | NP_077068.1 | 9 - 10 | 1410 | 121 | |
XM_006257378.2 | XP_006257440.1 | 7 - 8 | 1735 | 121 | ||
XM_006257381.2 | XP_006257443.1 | 9 - 10 | 1695 | 121 | ||
XM_008765793.1 | XP_008764015.1 | 7 - 8 | 1739 | 121 | ||
XM_008765794.1 | XP_008764016.1 | 9 - 10 | 1698 | 121 | ||
XM_008765795.1 | XP_008764017.1 | 9 - 10 | 1351 | 121 | ||
XM_008765796.1 | XP_008764018.1 | 9 - 10 | 1695 | 121 | ||
GenBank mRNA | AB049451.1 | - | 7 - 8 | 1001 | 121 | |
AJ006519.1 | - | 7 - 8 | 1456 | 121 | ||
AJ309926.1 | - | 7 - 8 | 1460 | 121 | ||
CO398759.1 | - | 2 - 3 | 155 | 121 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
