Assay Details
Gene Symbol: Sp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012655.2 | NP_036787.2 | 5 - 6 | 2091 | 78 | |
GenBank mRNA | D12768.1 | - | 5 - 6 | 2107 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
ossification
in utero embryonic development
trophectodermal cell differentiation
liver development
embryonic placenta development
regulation of transcription, DNA-templated
regulation of transcription by RNA polymerase II
transcription by RNA polymerase II
megakaryocyte differentiation
lung development
enucleate erythrocyte differentiation
positive regulation by host of viral transcription
positive regulation of transcription, DNA-templated
positive regulation of transcription by RNA polymerase II
rhythmic process
embryonic camera-type eye morphogenesis
embryonic skeletal system development
embryonic process involved in female pregnancy
definitive hemopoiesis
transcription cis-regulatory region binding
RNA polymerase II cis-regulatory region sequence-specific DNA binding
RNA polymerase II core promoter sequence-specific DNA binding
DNA-binding transcription factor activity, RNA polymerase II-specific
cis-regulatory region sequence-specific DNA binding
core promoter sequence-specific DNA binding
DNA binding
double-stranded DNA binding
DNA-binding transcription factor activity
protein binding
protein C-terminus binding
transcription factor binding
histone acetyltransferase binding
protein homodimerization activity
histone deacetylase binding
bHLH transcription factor binding
sequence-specific DNA binding
metal ion binding
RNA polymerase II-specific DNA-binding transcription factor binding
HMG box domain binding
ossification
in utero embryonic development
trophectodermal cell differentiation
liver development
embryonic placenta development
regulation of transcription, DNA-templated
regulation of transcription by RNA polymerase II
transcription by RNA polymerase II
megakaryocyte differentiation
lung development
enucleate erythrocyte differentiation
positive regulation by host of viral transcription
positive regulation of transcription, DNA-templated
positive regulation of transcription by RNA polymerase II
rhythmic process
embryonic camera-type eye morphogenesis
embryonic skeletal system development
embryonic process involved in female pregnancy
definitive hemopoiesis
transcription cis-regulatory region binding
RNA polymerase II cis-regulatory region sequence-specific DNA binding
RNA polymerase II core promoter sequence-specific DNA binding
DNA-binding transcription factor activity, RNA polymerase II-specific
cis-regulatory region sequence-specific DNA binding
core promoter sequence-specific DNA binding
DNA binding
double-stranded DNA binding
DNA-binding transcription factor activity
protein binding
protein C-terminus binding
transcription factor binding
histone acetyltransferase binding
protein homodimerization activity
histone deacetylase binding
bHLH transcription factor binding
sequence-specific DNA binding
metal ion binding
RNA polymerase II-specific DNA-binding transcription factor binding
HMG box domain binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P045418 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P045413 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 520284 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898772 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823527 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908006 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 320378 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732064 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106241 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574838 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 477906 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563764 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193986 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328063 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904218 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920378 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151924 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 85456 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 743585 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 410341 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 219907 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 454051 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211254 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251894 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115393 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 128480 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4957 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P045418 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P045413 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P045413 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P045418 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P045418 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P045413 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P454192 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 28907 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 94287 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 85501 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 65492 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10899553 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10899547 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10899551 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10899549 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10899546 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10899550 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10899548 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10899545 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10899552 | - | - |
Rat Genome 230 2.0 Array | 1392180_at | - | - |
Rat Genome 230 2.0 Array | 1392631_PM_at | - | - |
Rat Genome 230 2.0 Array | 1371880_at | - | - |
Rat Genome 230 2.0 Array | 1379384_PM_at | - | - |
Rat Genome 230 2.0 Array | 1379384_at | - | - |
Rat Genome 230 2.0 Array | 1369247_at | - | - |
Rat Genome 230 2.0 Array | 1369247_PM_at | - | - |
Rat Genome 230 2.0 Array | 1371880_PM_at | - | - |
Rat Genome 230 2.0 Array | 1392180_PM_at | - | - |
Rat Genome 230 2.0 Array | 1392631_at | - | - |
Affymetrix HT Rat Focus Array | 1392180_at | - | - |
Affymetrix HT Rat Focus Array | 1371880_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1379384_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1369247_at | - | - |
Affymetrix HT Rat Focus Array | 1371880_at | - | - |
Affymetrix HT Rat Focus Array | 1369247_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1392180_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1379384_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0700014903.rn.1 | TC0700001901.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700014906.rn.1 | TC0700001901.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700014905.rn.1 | TC0700001901.rn.1 | RTA TC |
Rat Expression Set 230 | 1369247_PM_at | - | - |
Rat Expression Set 230 | 1369247_at | - | - |
Rat Expression Set 230 | 1371880_at | - | - |
Rat Expression Set 230 | 1371880_PM_at | - | - |
Rat Expression Set 230 | 1379384_at | - | - |
Rat Expression Set 230 | 1392631_at | - | - |
Rat Expression Set 230 | 1392180_at | - | - |
Rat Expression Set 230 | 1379384_PM_at | - | - |
Rat Expression Set 230 | 1392180_PM_at | - | - |
Rat Expression Set 230 | 1392631_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10899552 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10899545 | - | - |
Rat Genome U34 Set | D12768_at | - | - |
Rat Genome U34 Set | rc_AA900254_at | - | - |
Rat Genome U34 Set | rc_AI045226_at | - | - |
Rat Genome U34 Set | rc_AI145910_at | - | - |
Rat Genome U34 Set | rc_AI137591_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22091746 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1373417 | - | - |
RatRef-12 Expression BeadChip | ILMN_52507 | - | - |
