Assay Details
Gene Symbol: Tmbim6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019381.2 | NP_062254.2 | 2 - 3 | 139 | 106 | |
XM_006257323.2 | XP_006257385.1 | 2 - 3 | 281 | 106 | ||
XM_008765670.1 | XP_008763892.1 | 3 - 4 | 239 | 106 | ||
GenBank mRNA | AY321320.1 | - | 4 - 5 | 277 | 106 | |
AY325130.1 | - | 4 - 5 | 277 | 106 | ||
AY325243.1 | - | 4 - 5 | 277 | 106 | ||
BC058478.1 | - | 2 - 3 | 145 | 106 | ||
FQ217052.1 | - | 2 - 3 | 138 | 106 | ||
FQ226858.1 | - | 2 - 3 | 168 | 106 | ||
FQ230608.1 | - | 2 - 3 | 168 | 106 | ||
FQ231817.1 | - | 2 - 3 | 146 | 106 | ||
X75855.1 | - | 2 - 3 | 171 | 106 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC058478.1, NM_019381.2, X75855.1, XM_006257323.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
