Assay Details
Gene Symbol: Tmbim6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019381.2 | NP_062254.2 | 7 - 8 | 595 | 62 | |
XM_006257323.2 | XP_006257385.1 | 7 - 8 | 737 | 62 | ||
XM_008765670.1 | XP_008763892.1 | 8 - 9 | 695 | 62 | ||
GenBank mRNA | AY321320.1 | - | 9 - 10 | 733 | 62 | |
AY325130.1 | - | 9 - 10 | 733 | 62 | ||
AY325243.1 | - | 9 - 10 | 733 | 62 | ||
BC058478.1 | - | 7 - 8 | 601 | 62 | ||
FQ217052.1 | - | 7 - 8 | 594 | 62 | ||
FQ218664.1 | - | 7 - 8 | 597 | 62 | ||
FQ226858.1 | - | 7 - 8 | 624 | 62 | ||
FQ229652.1 | - | 7 - 8 | 596 | 62 | ||
FQ230608.1 | - | 7 - 8 | 624 | 62 | ||
FQ231817.1 | - | 7 - 8 | 602 | 62 | ||
X75855.1 | - | 7 - 8 | 624 | 62 | ||
X75856.1 | - | 6 - 7 | 508 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
