Assay Details
Gene Symbol: Scaf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271170.1 | NP_001258099.1 | 5 - 6 | 450 | 141 | |
GenBank mRNA | CB547108.1 | - | 5 - 6 | 456 | 141 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
