Assay Details
Gene Symbol: Letmd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122781.3 | NP_001116253.3 | 3 - 4 | 424 | 125 | |
XM_006242354.2 | XP_006242416.1 | 3 - 4 | 487 | 125 | ||
GenBank mRNA | BC159426.1 | - | 3 - 4 | 405 | 125 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
