Assay Details
Gene Symbol: Setdb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271175.1 | NP_001258104.1 | 6 - 7 | 778 | 89 | |
XM_006232913.2 | XP_006232975.1 | 7 - 8 | 1168 | 89 | ||
XM_006232914.2 | XP_006232976.1 | 6 - 7 | 949 | 89 | ||
XM_006232916.2 | XP_006232978.1 | 2 - 3 | 145 | 89 | ||
XM_006232917.2 | XP_006232979.1 | 2 - 3 | 648 | 89 |
Gene Symbol: LOC100911180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003752173.3 | XP_003752221.2 | 2 - 3 | 89 | ||
XM_008773705.1 | XP_008771927.1 | 1 - 2 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
