Assay Details
Gene Symbol: Prkab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006233011.2 | XP_006233073.1 | 4 - 5 | 625 | 138 | |
XM_006233012.2 | XP_006233074.1 | 4 - 5 | 525 | 138 | ||
GenBank mRNA | BC078821.1 | - | 4 - 5 | 625 | 138 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
