Assay Details
Gene Symbol: mrpl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007696.1 | NP_001007697.1 | 1 - 2 | 164 | 55 | |
GenBank mRNA | BC082085.1 | - | 1 - 2 | 164 | 55 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
