Assay Details
Gene Symbol: Vangl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109584.1 | NP_001103054.1 | 2 - 3 | 274 | 157 | |
XM_006233054.2 | XP_006233116.1 | 2 - 3 | 315 | 157 | ||
XM_006233055.2 | XP_006233117.1 | 2 - 3 | 314 | 157 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001109584.1, XM_006233054.2, XM_006233055.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
