Assay Details
Gene Symbol: Prpf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108559.1 | NP_001102029.1 | 11 - 12 | 1691 | 117 | |
XM_006232968.2 | XP_006233030.1 | 11 - 12 | 1699 | 117 | ||
XM_006232969.2 | XP_006233031.1 | 11 - 12 | 3373 | 117 | ||
XM_006232970.1 | XP_006233032.1 | 11 - 12 | 1665 | 117 | ||
XM_008761313.1 | XP_008759535.1 | 11 - 12 | 1700 | 117 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
